siRNA Details
| Gene Symbol: Tuba3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009449.3 | NP_033475.1 | 4 | 574 | ||
| GenBank mRNA | BC050769.1 | - | 4 | 498 | 6771187 | |
| M13443.1 | - | 4 | 489 | |||
| Gene Symbol: Tuba3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009446.2 | NP_033472.1 | 4 | 522 | ||
| GenBank mRNA | AK132775.1 | - | 4 | 522 | ||
| BC050770.1 | - | 4 | 508 | 6774775 | ||
| BC089547.1 | - | 4 | 508 | 6771690 | ||
| M13442.1 | - | 4 | 510 | |||

Tuba3b
Genomic Map