siRNA Details
| Gene Symbol: Tuft1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001293728.1 | NP_001280657.1 | 11, 12 | 1127 | ||
| NM_011656.3 | NP_035786.1 | 12, 13 | 1202 | |||
| GenBank mRNA | AF047704.1 | - | 12, 13 | 1136 | ||
| AK132488.1 | - | 12, 13 | 1134 | |||
| AK171705.1 | - | 11, 12 | 1092 | |||
| BC019213.1 | - | 12, 13 | 1133 | 5064658 | ||

Tuft1
Genomic Map