siRNA Details
| Gene Symbol: Ube2l3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009456.2 | NP_033482.1 | 2 | 81 | ||
| GenBank mRNA | AJ130961.1 | - | 2 | 70 | ||
| AK152522.1 | - | 2 | 83 | |||
| AK165620.1 | - | 2 | 62 | |||
| AK167847.1 | - | 2 | 69 | |||
| BC049075.1 | - | 2 | 68 | |||
| BC057959.1 | - | 2 | 62 | |||
| BC093503.1 | - | 2 | 60 | 4216580 | ||
| BC106149.1 | - | 2 | 59 | 4502101 | ||
| X97042.1 | - | 2 | 81 | |||

Ube2l3
Genomic Map