siRNA Details
| Gene Symbol: Uba3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF077330.1 | - | 13 | 1033 | ||
| AK014433.1 | - | 14 | 1076 | |||
| AK032514.1 | - | 14 | 1267 | |||
| AK048148.1 | - | 14 | 1065 | |||
| AK159381.1 | - | 14 | 1153 | |||
| AK166655.1 | - | 13 | 1023 | |||
| AK168859.1 | - | 14 | 1068 | |||
| AY029181.1 | - | 13 | 1033 | |||
| BC002002.1 | - | 14 | 1053 | 3590282 | ||
| BC080776.1 | - | 13 | 986 | |||
| CF928750.1 | - | 3 | 166 | |||

Uba3
Genomic Map