siRNA Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019803.3 | NP_062777.2 | 5, 6 | 448 | ||
| GenBank mRNA | AF296657.1 | - | 5, 6 | 429 | ||
| AK136365.1 | - | 5, 6 | 435 | |||
| AK146554.1 | - | 5, 6 | 453 | |||
| AK154464.1 | - | 5, 6 | 469 | |||
| BC010321.1 | - | 5, 6 | 448 | 2651046 | ||
| U93241.1 | - | 4, 5 | 348 | |||

Ube2g2
Genomic Map