siRNA Details
| Gene Symbol: Ube3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033962.1 | NP_001029134.1 | 4 | 1805 | ||
| NM_011668.2 | NP_035798.2 | 6 | 1992 | |||
| NM_173010.3 | NP_766598.1 | 4 | 1807 | |||
| GenBank mRNA | AK029133.1 | - | 4 | 1808 | ||
| AK083795.1 | - | 4 | 1737 | |||
| AK220549.1 | - | 4 | 1705 | |||
| BC131658.1 | - | 4 | 1793 | 40110636 | ||
| BC131659.1 | - | 4 | 1793 | 40110637 | ||
| U82122.1 | - | 2 | 1242 | |||
| U96636.1 | - | 6 | 1539 | |||

Ube3a
Genomic Map