siRNA Details
| Gene Symbol: Usp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011669.3 | NP_035799.1 | 6 | 973 | ||
| GenBank mRNA | AF022792.1 | - | 3 | 242 | ||
| AF441835.1 | - | 5 | 634 | |||
| AK006739.1 | - | 6 | 1023 | |||
| AK035629.1 | - | 6 | 814 | |||
| AK167153.1 | - | 6 | 813 | |||
| BC049274.1 | - | 4 | 640 | 4500832 | ||
| BC055398.1 | - | 2 | 65 | |||
| BC068136.1 | - | 6 | 973 | 30363916 | ||

Usp12
Genomic Map