siRNA Details
| Gene Symbol: Uchl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011670.2 | NP_035800.2 | 7 | 658 | ||
| GenBank mRNA | AB025313.1 | - | 7 | 509 | ||
| AF172334.1 | - | 7 | 497 | |||
| AK004066.1 | - | 7 | 535 | |||
| AK013729.1 | - | 7 | 538 | |||
| AK170728.1 | - | 5 | 496 | |||
| BC039177.1 | - | 7 | 524 | 4506229 | ||
| CX241178.1 | - | 7 | 659 | |||

Uchl1
Genomic Map