siRNA Details
| Gene Symbol: Usp10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009462.1 | NP_033488.1 | 10 | 1739 | ||
| GenBank mRNA | AK077453.1 | - | 10 | 1761 | ||
| AK129083.1 | - | 10 | 1747 | |||
| AK137015.1 | - | 10 | 1761 | |||
| AK153675.1 | - | 10 | 2231 | |||
| AK157970.1 | - | 10 | 1763 | |||
| AK165156.1 | - | 10 | 1759 | |||
| AK166243.1 | - | 10 | 1755 | |||
| AK172443.1 | - | 10 | 1761 | |||
| BC007134.1 | - | 10 | 1741 | 3491297 | ||
| D84096.1 | - | 10 | 1739 | |||

Usp10
Genomic Map