siRNA Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009464.3 | NP_033490.1 | 4 | 672 | ||
| GenBank mRNA | AB008216.1 | - | 3 | 475 | ||
| AB010742.1 | - | 4 | 672 | |||
| AB013132.1 | - | 2 | 228 | |||
| AF030164.1 | - | 3 | 513 | |||
| AF032902.1 | - | 4 | 639 | |||
| AF053352.1 | - | 4 | 644 | |||
| BC055901.1 | - | 3 | 862 | |||
| BC098234.1 | - | 2 | 1368 | |||
| BC139430.1 | - | 3 | 540 | 8862452 | ||
| BC139431.1 | - | 3 | 540 | 8862453 | ||

Ucp3
Genomic Map