siRNA Details
| Gene Symbol: Ucp3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009464.3 | NP_033490.1 | 7 | 1067 | ||
| GenBank mRNA | AB008216.1 | - | 6 | 870 | ||
| AB010742.1 | - | 7 | 1067 | |||
| AF030164.1 | - | 6 | 908 | |||
| AF032902.1 | - | 7 | 1034 | |||
| AF053352.1 | - | 7 | 1039 | |||
| BC055901.1 | - | 6 | 1257 | |||
| BC098234.1 | - | 5 | 1763 | |||
| BC139430.1 | - | 6 | 935 | 8862452 | ||
| BC139431.1 | - | 6 | 935 | 8862453 | ||

Ucp3
Genomic Map