siRNA Details
| Gene Symbol: Ufd1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011672.4 | NP_035802.3 | 5 | 528 | ||
| GenBank mRNA | AK010461.1 | - | 5 | 497 | ||
| AK012543.1 | - | 3 | 472 | |||
| AK088823.1 | - | 5 | 497 | |||
| AK147138.1 | - | 5 | 511 | |||
| AK169062.1 | - | 5 | 510 | |||
| BC006630.1 | - | 5 | 485 | 3708320 | ||
| U64445.1 | - | 5 | 513 | |||

Ufd1l
Genomic Map