siRNA Details
| Gene Symbol: Dpysl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136086.2 | NP_001129558.1 | 9 | 1027 | ||
| NM_001291455.1 | NP_001278384.1 | 9 | 1277 | |||
| NM_009468.5 | NP_033494.1 | 9 | 1208 | |||
| GenBank mRNA | AK077000.1 | - | 9 | 1018 | ||
| AK133040.1 | - | 9 | 1027 | |||
| AK149024.1 | - | 9 | 1046 | |||
| AK161509.1 | - | 9 | 949 | |||
| AK171653.1 | - | 2 | 108 | |||
| BC023003.1 | - | 9 | 892 | 5367381 | ||
| X87817.1 | - | 9 | 1168 | |||

Dpysl3
Genomic Map