siRNA Details
| Gene Symbol: Ulk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009469.3 | NP_033495.2 | 4, 5 | 641 | ||
| GenBank mRNA | AF053756.1 | - | 4, 5 | 333 | ||
| AF072370.1 | - | 4, 5 | 324 | |||
| AK122357.1 | - | 3, 4 | 143 | |||
| AK148317.1 | - | 4, 5 | 616 | |||
| AK154939.1 | - | 4, 5 | 658 | |||
| AK156534.1 | - | 4, 5 | 618 | |||
| BC053062.1 | - | 4, 5 | 301 | |||
| BC057121.1 | - | 4, 5 | 641 | 6834534 | ||
| BC059835.1 | - | 4, 5 | 592 | 6406755 | ||

Ulk1
Genomic Map