siRNA Details
| Gene Symbol: Umod | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278605.1 | NP_001265534.1 | 8 | 1877 | ||
| NM_009470.5 | NP_033496.1 | 8 | 1933 | |||
| GenBank mRNA | AK085460.1 | - | 8 | 1927 | ||
| AK144065.1 | - | 8 | 1849 | |||
| AK164688.1 | - | 8 | 1849 | |||
| AK165507.1 | - | 8 | 1793 | |||
| BC012973.1 | - | 7 | 1771 | 4210940 | ||
| L33406.1 | - | 8 | 1863 | |||

Umod
Genomic Map