siRNA Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009471.2 | NP_033497.1 | 2 | 309 | ||
| GenBank mRNA | AK035821.1 | - | 2 | 310 | ||
| AK090102.1 | - | 2 | 312 | |||
| BC003887.1 | - | 2 | 306 | 3590810 | ||
| BC095981.1 | - | 2 | 295 | 3499037 | ||

Umps
Genomic Map