siRNA Details
| Gene Symbol: Umps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009471.2 | NP_033497.1 | 5 | 1244 | ||
| GenBank mRNA | AK035821.1 | - | 5 | 1245 | ||
| AK035997.1 | - | 4 | 1092 | |||
| AK090102.1 | - | 5 | 1250 | |||
| AK159808.1 | - | 5 | 1245 | |||
| AK159955.1 | - | 5 | 1245 | |||
| AK183780.1 | - | 2 | 194 | |||
| BC003887.1 | - | 5 | 1241 | 3590810 | ||
| BC095981.1 | - | 5 | 1230 | 3499037 | ||
| M29395.1 | - | 3 | 577 | |||

Umps
Genomic Map