siRNA Details
| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009474.5 | NP_033500.1 | 2 | 100 | ||
| GenBank mRNA | AK050425.1 | - | 2 | 101 | ||
| BC019771.1 | - | 2 | 82 | 5136328 | ||
| M27695.1 | - | 2 | 55 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Uox| Gene Symbol: Uox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009474.5 | NP_033500.1 | 2 | 100 | ||
| GenBank mRNA | AK050425.1 | - | 2 | 101 | ||
| BC019771.1 | - | 2 | 82 | 5136328 | ||
| M27695.1 | - | 2 | 55 | |||