siRNA Details
| Gene Symbol: ITGB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000212.2 | NP_000203.2 | 13 | 2065 | ||
| GenBank mRNA | BC127666.1 | - | 13 | 2065 | 40128462 | |
| BC127667.1 | - | 13 | 2069 | 40128464 | ||
| J02703.1 | - | 13 | 2065 | |||
| KC120777.1 | - | 13 | 2066 | |||
| KC120778.1 | - | 13 | 2065 | |||
| M20311.1 | - | 13 | 2061 | |||
| M35999.1 | - | 13 | 2063 | |||
| U95204.1 | - | 13 | 2045 | |||

ITGB3
Genomic Map