siRNA Details
| Gene Symbol: Uros | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AA272198.1 | - | 5 | 485 | ||
| AK135314.1 | - | 6 | 541 | |||
| AK145902.1 | - | 6 | 591 | |||
| AK148116.1 | - | 6 | 728 | |||
| AK164297.1 | - | 6 | 506 | |||
| BC068161.1 | - | 6 | 534 | 30358504 | ||
| U04439.1 | - | 5 | 455 | |||
| U16216.1 | - | 6 | 596 | |||
| U18867.2 | - | 6 | 600 | |||

Uros
Genomic Map