siRNA Details
| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF479773.1 | - | 7 | 612 | ||
| AK142437.1 | - | 9 | 865 | |||
| BC049784.1 | - | 9 | 808 | 6506829 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usf1| Gene Symbol: Usf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF479773.1 | - | 7 | 612 | ||
| AK142437.1 | - | 9 | 865 | |||
| BC049784.1 | - | 9 | 808 | 6506829 | ||