siRNA Details
| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009481.2 | NP_033507.2 | 25 | 4159 | ||
| GenBank mRNA | DQ086491.1 | - | 25 | 4120 | ||
| U67874.1 | - | 25 | 4120 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Usp9x| Gene Symbol: Usp9x | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009481.2 | NP_033507.2 | 25 | 4159 | ||
| GenBank mRNA | DQ086491.1 | - | 25 | 4120 | ||
| U67874.1 | - | 25 | 4120 | |||