siRNA Details
| Gene Symbol: Kdm6a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009483.1 | NP_033509.1 | 3 | 585 | ||
| GenBank mRNA | AJ002730.1 | - | 2 | 44 | ||
| AK080370.1 | - | 3 | 604 | |||
| AK144654.1 | - | 3 | 611 | |||
| AK164941.1 | - | 3 | 617 | |||
| BC053433.1 | - | 3 | 585 | 6516368 | ||

Kdm6a
Genomic Map