siRNA Details
| Gene Symbol: Uty | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009484.2 | NP_033510.2 | 15 | 1915 | ||
| GenBank mRNA | AF057367.1 | - | 15 | 1895 | ||
| AK142344.1 | - | 18 | 2053 | |||
| AK142512.1 | - | 15 | 1831 | |||
| AK155922.1 | - | 15 | 1660 | |||
| AK162410.1 | - | 13 | 1488 | |||
| AK164380.1 | - | 1 | 1852 | |||
| AK164917.1 | - | 2 | 360 | |||
| AK220463.1 | - | 16 | 1872 | |||
| BC053061.1 | - | 15 | 1898 | |||

Uty
Genomic Map