siRNA Details
| Gene Symbol: ITGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000213.4 | NP_000204.3 | 5 | 719 | ||
| NM_001005619.1 | NP_001005619.1 | 4 | 432 | |||
| NM_001005731.2 | NP_001005731.1 | 5 | 719 | |||
| NM_001321123.1 | NP_001308052.1 | 5 | 670 | |||
| XM_005257309.2 | XP_005257366.1 | 5 | 489 | |||
| XM_005257311.4 | XP_005257368.1 | 5 | 708 | |||
| XM_006721866.3 | XP_006721929.1 | 5 | 635 | |||
| XM_006721867.3 | XP_006721930.1 | 5 | 632 | |||
| XM_006721868.3 | XP_006721931.1 | 5 | 629 | |||
| XM_006721870.3 | XP_006721933.1 | 5 | 625 | |||
| XM_011524751.2 | XP_011523053.1 | 5 | 633 | |||
| GenBank mRNA | AB208913.1 | - | 5 | 521 | ||
| AF011375.1 | - | 4 | 424 | |||
| AK304792.1 | - | 5 | 489 | |||
| AK310952.1 | - | 5 | 489 | |||
| BC109238.1 | - | 1 | 177 | |||
| BC118916.1 | - | 5 | 481 | 40073279 | ||
| BC126411.1 | - | 5 | 540 | 8992127 | ||
| BC143738.1 | - | 5 | 540 | 9052252 | ||
| BC143742.1 | - | 5 | 491 | 9052256 | ||
| X51841.1 | - | 5 | 550 | |||
| X52186.1 | - | 4 | 433 | |||
| X53587.1 | - | 5 | 611 | |||

ITGB4
Genomic Map