siRNA Details
| Gene Symbol: Vmn1r51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011683.2 | NP_035813.2 | 3 | 1708 | ||
| GenBank mRNA | BC046308.1 | - | 3 | 1708 | 6306893 | |
| Y12725.1 | - | 2 | 1678 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r51| Gene Symbol: Vmn1r51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011683.2 | NP_035813.2 | 3 | 1708 | ||
| GenBank mRNA | BC046308.1 | - | 3 | 1708 | 6306893 | |
| Y12725.1 | - | 2 | 1678 | |||