siRNA Details
| Gene Symbol: Vmn2r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009491.3 | NP_033517.3 | 3 | 1149 | ||
| GenBank mRNA | AF011426.1 | - | 901 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r10| Gene Symbol: Vmn2r10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009491.3 | NP_033517.3 | 3 | 1149 | ||
| GenBank mRNA | AF011426.1 | - | 901 | |||