siRNA Details
| Gene Symbol: Vmn2r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009493.2 | NP_033519.1 | 3 | 915 | ||
| GenBank mRNA | AF011414.1 | - | 3 | 915 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn2r42| Gene Symbol: Vmn2r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009493.2 | NP_033519.1 | 3 | 915 | ||
| GenBank mRNA | AF011414.1 | - | 3 | 915 | ||