siRNA Details
| Gene Symbol: Vmn2r42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009493.2 | NP_033519.1 | 2 | 493 | ||
| GenBank mRNA | AF011414.1 | - | 2 | 493 | ||
| Gene Symbol: Vmn2r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105151.1 | NP_001098621.1 | 2 | 377 | ||
More Information
Set Membership: |
|

Vmn2r42
Genomic Map