siRNA Details
| Gene Symbol: Vars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011690.3 | NP_035820.3 | 4 | 925 | ||
| GenBank mRNA | AF087680.1 | - | 4 | 688 | ||
| AK012123.1 | - | 4 | 750 | |||
| AK154823.1 | - | 4 | 740 | |||
| AK157814.1 | - | 4 | 783 | |||
| AK165326.1 | - | 3 | 368 | |||
| BC053703.1 | - | 4 | 708 | 30055190 | ||

Vars
Genomic Map