siRNA Details
| Gene Symbol: Vars | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011690.3 | NP_035820.3 | 29, 30 | 4081 | ||
| GenBank mRNA | AF087680.1 | - | 29, 30 | 3844 | ||
| AK019157.1 | - | 21, 22 | 2478 | |||
| AK154823.1 | - | 29, 30 | 3896 | |||
| AK165326.1 | - | 27, 28 | 3596 | |||
| AK167842.1 | - | 21, 22 | 2619 | |||
| BB070705.1 | - | 1, 2 | 111 | |||
| BC053703.1 | - | 29, 30 | 3864 | 30055190 | ||

Vars
Genomic Map