siRNA Details
| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009502.4 | NP_033528.3 | 13 | 1895 | ||
| GenBank mRNA | AK077850.1 | - | 13 | 1931 | ||
| BC008520.1 | - | 13 | 1895 | 3495710 | ||
| L18880.1 | - | 13 | 1824 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vcl| Gene Symbol: Vcl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009502.4 | NP_033528.3 | 13 | 1895 | ||
| GenBank mRNA | AK077850.1 | - | 13 | 1931 | ||
| BC008520.1 | - | 13 | 1895 | 3495710 | ||
| L18880.1 | - | 13 | 1824 | |||