siRNA Details
| Gene Symbol: ITGB5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002213.4 | NP_002204.2 | 8 | 1373 | ||
| XM_006713630.2 | XP_006713693.1 | 8 | 1606 | |||
| XM_017006351.1 | XP_016861840.1 | 8 | 1144 | |||
| XM_017006352.1 | XP_016861841.1 | 8 | 1083 | |||
| XM_017006353.1 | XP_016861842.1 | 8 | 1115 | |||
| GenBank mRNA | AK315429.1 | - | 8 | 1109 | ||
| BC006541.2 | - | 8 | 1365 | 2958666 | ||
| J05633.1 | - | 8 | 1114 | |||
| M35011.1 | - | 8 | 1097 | |||
| X53002.1 | - | 8 | 1404 | |||

ITGB5
Genomic Map