siRNA Details
| Gene Symbol: ITGB6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_000888.4 | NP_000879.2 | 7 | 1236 | ||
| NM_001282353.1 | NP_001269282.1 | 8 | 1129 | |||
| NM_001282354.1 | NP_001269283.1 | 5 | 951 | |||
| NM_001282355.1 | NP_001269284.1 | 7 | 1236 | |||
| NM_001282388.1 | NP_001269317.1 | 6 | 1156 | |||
| NM_001282389.1 | NP_001269318.1 | 7 | 1063 | |||
| NM_001282390.1 | NP_001269319.1 | 6 | 983 | |||
| GenBank mRNA | AK290300.1 | - | 7 | 1204 | ||
| AK304203.1 | - | 6 | 1095 | |||
| AK313944.1 | - | 8 | 1120 | |||
| AL831998.1 | - | 7 | 1175 | |||
| BC121178.2 | - | 7 | 1006 | 40122759 | ||
| BC143656.1 | - | 7 | 1014 | |||
| CD013944.1 | - | 6 | 928 | |||
| CD013945.1 | - | 7 | 1181 | |||
| CD013946.1 | - | 5 | 896 | |||
| CD013947.1 | - | 7 | 1008 | |||
| M35198.3 | - | 7 | 1002 | |||

ITGB6
Genomic Map