siRNA Details
| Gene Symbol: Vezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016686.4 | NP_057895.2 | 2 | 254 | ||
| GenBank mRNA | AF104410.3 | - | 2 | 254 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vezf1| Gene Symbol: Vezf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_016686.4 | NP_057895.2 | 2 | 254 | ||
| GenBank mRNA | AF104410.3 | - | 2 | 254 | ||