siRNA Details
| Gene Symbol: Vipr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009511.2 | NP_033537.1 | 2 | 202 | ||
| GenBank mRNA | AK038743.1 | - | 2 | 203 | ||
| AK142357.1 | - | 2 | 168 | |||
| BC138572.1 | - | 2 | 100 | 8861592 | ||
| D28132.1 | - | 2 | 134 | |||
More Information
Set Membership: |
|

Vipr2
Genomic Map