siRNA Details
| Gene Symbol: Vldlr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161420.1 | NP_001154892.1 | 11 | 2282 | ||
| NM_013703.2 | NP_038731.2 | 11 | 2282 | |||
| GenBank mRNA | AK044683.1 | - | 3 | 890 | ||
| AK076172.1 | - | 11 | 2287 | |||
| AK136838.1 | - | 11 | 2102 | |||
| BC013622.1 | - | 11 | 1718 | 3968213 | ||
| L33417.1 | - | 11 | 1706 | |||
| U06670.1 | - | 11 | 1868 | |||

Vldlr
Genomic Map