siRNA Details
| Gene Symbol: ITIH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002215.3 | NP_002206.2 | 1 | 76 | ||
| GenBank mRNA | AK292750.1 | - | 1 | 46 | ||
| AK298398.1 | - | 1 | 75 | |||
| AK298423.1 | - | 1 | 68 | |||
| AK314198.1 | - | 1 | 46 | |||
| BC069464.1 | - | 1 | 70 | 7262464 | ||
| BX496392.1 | - | 1 | 77 | |||
| X63652.1 | - | 1 | 70 | |||

ITIH1
Genomic Map