siRNA Details
| Gene Symbol: Wbp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011712.2 | NP_035842.1 | 3 | 517 | ||
| GenBank mRNA | AK002214.1 | - | 3 | 471 | ||
| AK017717.1 | - | 3 | 477 | |||
| AK213578.1 | - | 1 | 124 | |||
| AY459189.1 | - | 3 | 516 | |||
| BC007478.1 | - | 3 | 414 | 3490650 | ||
| BC020289.1 | - | 3 | 442 | |||
| BU511227.1 | - | 3 | 548 | |||
| U92454.1 | - | 1 | 412 | |||

Wbp5
Genomic Map