siRNA Details
| Gene Symbol: Wee1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009516.3 | NP_033542.2 | 7 | 1641 | ||
| GenBank mRNA | AK011212.1 | - | 7 | 1417 | ||
| AK039572.1 | - | 7 | 957 | |||
| AK184246.1 | - | 2 | 120 | |||
| AK215663.1 | - | 2 | 120 | |||
| BC006852.1 | - | 7 | 1660 | 3156953 | ||

Wee1
Genomic Map