siRNA Details
| Gene Symbol: Wfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011716.2 | NP_035846.1 | 7 | 849 | ||
| GenBank mRNA | AF084482.1 | - | 7 | 782 | ||
| AJ011971.1 | - | 7 | 776 | |||
| AK144561.1 | - | 7 | 801 | |||
| AK145023.1 | - | 7 | 821 | |||
| AK170180.1 | - | 7 | 804 | |||
| AK185636.1 | - | 3 | 272 | |||
| AK189923.1 | - | 3 | 270 | |||
| BC046988.1 | - | 7 | 785 | 6492038 | ||
| CX562372.1 | - | 5 | 479 | |||

Wfs1
Genomic Map