siRNA Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 3 | 511 | ||
| GenBank mRNA | AF012923.1 | - | 3 | 465 | ||
| BC138740.1 | - | 2 | 330 | 8861762 | ||
| BC138742.1 | - | 2 | 330 | 8861764 | ||
| BQ895699.1 | - | 3 | 524 | |||
| CD804474.1 | - | 1 | 14 | |||

Zmat3
Genomic Map