siRNA Details
| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 5 | 856 | ||
| GenBank mRNA | BC138740.1 | - | 4 | 675 | 8861762 | |
| BC138742.1 | - | 4 | 675 | 8861764 | ||
| CD804474.1 | - | 3 | 359 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zmat3| Gene Symbol: Zmat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009517.2 | NP_033543.2 | 5 | 856 | ||
| GenBank mRNA | BC138740.1 | - | 4 | 675 | 8861762 | |
| BC138742.1 | - | 4 | 675 | 8861764 | ||
| CD804474.1 | - | 3 | 359 | |||