siRNA Details
| Gene Symbol: Wnt3a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009522.2 | NP_033548.1 | 2 | 282 | ||
| GenBank mRNA | X56842.1 | - | 2 | 306 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001107005, XM_006246487.

Wnt3a
Genomic Map