siRNA Details
| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 4 | 455 | ||
| GenBank mRNA | AF130349.1 | - | 4 | 457 | ||
| AK050992.1 | - | 4 | 456 | |||
| BC119544.2 | - | 4 | 387 | 40056929 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Wnt8b| Gene Symbol: Wnt8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011720.3 | NP_035850.2 | 4 | 455 | ||
| GenBank mRNA | AF130349.1 | - | 4 | 457 | ||
| AK050992.1 | - | 4 | 456 | |||
| BC119544.2 | - | 4 | 387 | 40056929 | ||