siRNA Details
| Gene Symbol: Wrn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122822.1 | NP_001116294.1 | 21 | 2658 | ||
| NM_011721.4 | NP_035851.3 | 21 | 2768 | |||
| GenBank mRNA | AF091215.1 | - | 21 | 2722 | ||
| AF241636.1 | - | 21 | 2783 | |||
| AK143778.1 | - | 20 | 2597 | |||
| AK161483.1 | - | 15 | 2553 | |||
| AK171490.1 | - | 21 | 2769 | |||
| BC050921.1 | - | 21 | 2713 | 6408901 | ||
| BC060700.1 | - | 21 | 2713 | 6408901 | ||
| D86526.1 | - | 21 | 2738 | |||
| D86527.1 | - | 21 | 2738 | |||

Wrn
Genomic Map