siRNA Details
| Gene Symbol: Xdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011723.3 | NP_035853.2 | 3 | 240 | ||
| GenBank mRNA | AK034256.1 | - | 3 | 190 | ||
| AK143412.1 | - | 3 | 189 | |||
| AK144700.1 | - | 3 | 192 | |||
| AK161529.1 | - | 3 | 191 | |||
| AK164764.1 | - | 3 | 183 | |||
| BC141183.1 | - | 3 | 215 | 9055795 | ||
| BY322557.1 | - | 3 | 226 | |||
| X62932.1 | - | 3 | 174 | |||

Xdh
Genomic Map