siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 7 | 748 | ||
| GenBank mRNA | AF026032.1 | - | 7, 8 | 715 | ||
| AK032955.1 | - | 2, 3 | 160 | |||
| AK088095.1 | - | 9 | 1143 | |||
| AK135019.1 | - | 7 | 905 | |||

Atrx
Genomic Map