siRNA Details
| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 14 | 4499 | ||
| GenBank mRNA | AF026032.1 | - | 15 | 4466 | ||
| AK134024.1 | - | 6 | 624 | |||
| AK161784.1 | - | 3 | 245 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Atrx| Gene Symbol: Atrx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_009530.2 | NP_033556.2 | 14 | 4499 | ||
| GenBank mRNA | AF026032.1 | - | 15 | 4466 | ||
| AK134024.1 | - | 6 | 624 | |||
| AK161784.1 | - | 3 | 245 | |||