siRNA Details
| Gene Symbol: Xpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011728.2 | NP_035858.2 | 4 | 507 | ||
| GenBank mRNA | AF354052.1 | - | 2 | 122 | ||
| AK008962.1 | - | 4 | 501 | |||
| BC029241.1 | - | 484 | 4489063 | |||
| CB058423.1 | - | 4 | 509 | |||
| X74351.1 | - | 4 | 507 | |||

Xpa
Genomic Map